Drukuj
Filtry do fazy ruchomej Last Drop
Dostawca: VICI JOUR RESEARCH
JR-9000-0530 JR-9000-0520 JR-9000-0521 JR-9000-0522 JR-9000-0531 JR-9000-0532 JR-9000-0532F JR-9000-0531F JR-9000-0530F JR-9000-0522F JR-9000-0520F JR-9000-0521F
554-1115EA 286 PLN
554-1115 554-1112 554-1113 554-1114 554-1116 554-1117 JOURJR-9000-0532F JOURJR-9000-0531F JOURJR-9000-0530F JOURJR-9000-0522F JOURJR-9000-0520F JOURJR-9000-0521F
Filtry do fazy ruchomej Last Drop
Filtry Filtry do fazy ruchomej
With flat filter element which allows the maximum amount of mobile phase to be drawn from the reservoir without introducing air into the system.