Drukuj
Filtry rozdzielające fazy
Dostawca: PRAT DUMAS
A090108H A110108H A125108H A150108H A185108H A210108H A240108H A270108H
PRATA090108HEA 282 PLN
PRATA090108H PRATA110108H PRATA125108H PRATA150108H PRATA185108H PRATA210108H PRATA240108H PRATA270108H
Filtry rozdzielające fazy
Filtry Sączki Sączki ekstrakcyjne (separator fazy)
Hydrophobic paper for the fast separation of aqueous solutions from hydrofuge solvents.
Useful as well with organic solvents heavier or lighter than water replacing many times bulbs to be settled.
Dodatkowe informacje: Filters supplied as flat in boxes with 100 filters or folded in 4 by 100.