Drukuj

Anti-SYT Mouse Monoclonal Antibody

Dostawca: ENZO LIFE SCIENCES
ADISYA130D ADISYA130F
ENZOADISYA130DEA 1380 PLN
ENZOADISYA130D ENZOADISYA130F
Anti-SYT Mouse Monoclonal Antibody
Przeciwciała
Synaptotagmins (Syt) are membrane-trafficking proteins characterized by an N-terminal transmembrane region, a variable linker, and two C-terminal C2-domains, C2A and C2B. At least twelve Synaptotagamins are expressed in vertebrates and can be found in both vesicular and plasma membranes.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse and rat Synaptotagmin. Detects a band of ~65kDa by Western blot.

Type: Primary
Antigen: SYT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR