Drukuj
Anti-CRYAB Rabbit Polyclonal Antibody
Dostawca: ENZO LIFE SCIENCES
ADISPA226D ADISPA226F
ENZOADISPA226DEA 1530 PLN
ENZOADISPA226D ENZOADISPA226F
Anti-CRYAB Rabbit Polyclonal Antibody
Przeciwciała
Alpha-crystallins, which are part of the small Heat shock family members, are major water-soluble proteins present in the lens of the mammalian eye. Phosphorylation of serine residues which occurs during development and in response to stress, is intimately linked with its function. Chaperone activity requires, and is modulated by, oligomerization and is limited to binding unfolded intermediates to prevent irreversible aggregation.
  • For Immunoprecipitation, Western Blot
Recognizes human, mouse, rat, bovine, chicken, dog, guinea pig, hamster, monkey, pig, rabbit, scallop and Xenopus alphaB-crystallin. Detects a band of ~22kDa by Western blot.
Type: Primary
Antigen: CRYAB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Zobacz ceny