Drukuj
Anti-CUL2 Mouse Monoclonal Antibody
Dostawca: ENZO LIFE SCIENCES
ADIKAMCC135D ADIKAMCC135F
ENZOADIKAMCC135DEA 1420 PLN
ENZOADIKAMCC135D ENZOADIKAMCC135F
Anti-CUL2 Mouse Monoclonal Antibody
Przeciwciała
The human Cullin family of genes consists of Cul-1, Cul-2, Cul-3, Cul-4a, Cul-4b, and Cul-5. Among the 6 members, Cul-1 and Cul-2 are very well characterized. Cul-2 is homologous to the Saccharomyces cerevisiae cdc53 gene. Studies have shown that Cul-1 forms an E3 ubiquitin-ligase, called the SCF complex (Skp1 and F-box protein such as bTrCp or FWD1), which controls the proteolysis of G1 cell cycle regulatory proteins.
  • For Western Blot
Recognizes human, mouse, rat, bovine, monkey and rabbit Cullin 1. Detects a band of ~93kDa by Western blot.
Type: Primary
Antigen: CUL2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Zobacz ceny