Drukuj

Anti-HSP90A Rabbit Polyclonal Antibody

Dostawca: ENZO LIFE SCIENCES
ADISPS771D ADISPS771F
ENZOADISPS771DEA 1130 PLN
ENZOADISPS771D ENZOADISPS771F
Anti-HSP90A Rabbit Polyclonal Antibody
Przeciwciała
The Hsp90 family of heat shock proteins represents one of the most abundantly expressed and highly conserved families of cellular chaperones whose expression can be upregulated under conditions of cellular stress, and includes cytoplasmic (Hsp90-alpha/beta), ER (grp94), and mitochondrial (TRAP1) localized members.

  • Dilution range: Western blot (1:1,000, colorimetric)
  • Detects a band of ~90kDa by Western blot

Type: Primary
Antigen: HSP90A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Beluga, Bovine, Dog, Fish, Guinea pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR