Drukuj
Standardowe głowice pomp Masterflex® L/S® do rurek precyzyjnych, Avantor®
Dostawca: Avantor Fluid Handling
MFLX07013-20EA 1440 PLN
MFLX07013-20 MFLX07013-21 MFLX07013-52 MFLX07014-20 MFLX07014-21 MFLX07014-52 MFLX07016-20 MFLX07016-21 MFLX07016-52 MFLX07017-20 MFLX07017-21 MFLX07017-52 MFLX07018-20 MFLX07018-21 MFLX07018-52
Standardowe głowice pomp Masterflex® L/S® do rurek precyzyjnych, Avantor®
Pompy Głowice pomp Głowice pomp perystaltycznych
Ideal for fluid transfer applications requiring high accuracy and repeatability.
  • Standard heads are precision molded to accommodate a single size of tubing
  • Pump head is compact and simple in design with same side tubing entry/exit—perfect for space-limited applications
  • Standard heads are compact and easy to integrate into your OEM applications
  • Stack up to four heads depending on drive recommendations
This Standard pump head accepts precision tubing— ensures optimal performance from your Masterflex pump. Select a pump head for one of the followingWięcej informacji o produkcie