Drukuj
Masterflex® L/S® Standard Pump Head for Precision Tubing L/S® 13, Polycarbonate Housing, SS Rotor
Dostawca: Avantor Fluid Handling
VWR Item#MFLX07013-21
Your Organization's Price:
Ideal for fluid transfer applications requiring high accuracy and repeatability.
  • Standard heads are precision molded to accommodate a single size of tubing
  • Pump head is compact and simple in design with same side tubing entry/exit—perfect for space-limited applications
  • Standard heads are compact and easy to integrate into your OEM applications
  • Stack up to four heads depending on drive recommendations
  • Rotor is constructed of stainless steel (SS) for enhanced durability
  • Suitable for use with L/S® 13 Precision pump tubing
This Standard pump head accepts precision tubing— ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 13, L/S 14, L/S 16, L/S 17, L/S 18.
<b>Dodatkowe informacje: </b>Includes tubing loading key and single-channel mounting hardware.