Drukuj
Zestaw ELISA do oznaczania proinsuliny, Grow Cells
Dostawca: ENZO LIFE SCIENCES
ENZKIT1490001
ENZOENZKIT1490001EA 3680 PLN
ENZOENZKIT1490001
Zestaw ELISA do oznaczania proinsuliny, Grow Cells
Zestawy do oznaczeń life science Testy ELISA
Ultra-sensitive Proinsulin ELISA kit enabling detection of proinsulin in human serum and plasma samples in just 3 hours.
  • Ultra-sensitive measurement of proinsulin, detecting as little as 0,17 pM
  • Negligible cross reactivity with insulin
  • High throughput format with results in three hours for up to 40 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of proinsulin in human serum and plasma samples.The Proinsulin ELISA kit is a colorimetric immunometric immunoassay kit with results in three hours.
Informacje do zamówienia: Contains Microtiter plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.