Zestaw ELISA do oznaczania CRP (ludzki)
Dostawca: ENZO LIFE SCIENCES
Sensitive (12,685ng/ml) ELISA kit for the quantification of CRP, an early biomarker for inflammation
- Sensitive measurement of CRP, detecting as little as 12,685 ng/ml
- Negligible reactivity for similar family members
- High throughput format with results in <2 hours for up to 40 samples in duplicate
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of human C-reactive protein in serum and plasma.
The CRP (human) ELISA kit is a complete, colorimetric, competitive immunoassay kit for the quantitative determination of human CRP in serum and plasma with results in less than two hours.
Informacje do zamówienia: Contains Microtiter Plate, Assay Buffer, Standard, Conjugate, Antibody, Streptavidin-HRP, TMB Substrate, Stop Solution, Wash Buffer Concentrate.