Drukuj
Organelle-ID™ RGB reagent II
Dostawca: ENZO LIFE SCIENCES
ENZ53008C200
ENZOENZ53008C200EA 2540 PLN
ENZOENZ53008C200
Organelle-ID™ RGB reagent II
Systemy do etykietowania i łączenia białek
Mixture of fluorescent dyes for detection of lysosomes, endoplasmic reticulum and nucleic acids
The Organelle-ID™ RGB reagent II is a mixture of cell-permeable red fluorescent lysosomal dye, green fluorescent endoplasmic reticulum dye and blue fluorescent nucleic acid dye. The staining pattern arising from the combination of these three dyes permits visualization of the target organelles by fluorescence/confocal microscopy. The reagent, supplied as a 500X solution, is sufficient for 1000 microscopy assays. The single-tube format makes this multi-organelle stain reagent easy to use.
Informacje do zamówienia: Quantity: 200 μl (for 1000 microscopy assays).