Drukuj
SAMS Peptide
Dostawca: ENZO LIFE SCIENCES
BMLP2310001
ENZOBMLP2310001EA 810 PLN
ENZOBMLP2310001
SAMS Peptide
Białka
AMPK substrate
This peptide, HMRSAMSGLHLVKRR, is derived from rat acetyl-CoA carboxylase and is known to be a good substrate for AMPK.