Drukuj
IKK substrate
Dostawca: ENZO LIFE SCIENCES
BMLP2260001
ENZOBMLP2260001EA 1170 PLN
ENZOBMLP2260001
IKK substrate
Białka
This peptide, KKKKERLLDDRHDSGLDSMKDEE, was derived from the IkappaBalpha sequence.
The four lysines on the amino-terminus allow this peptide to be used in phosphocellulose kinase assays for IKK activity.