Drukuj
PKI
Dostawca: ENZO LIFE SCIENCES
BMLP2030500
ENZOBMLP2030500EA 695 PLN
ENZOBMLP2030500
PKI
Białka
PKA inhibitor
Sequence is from the heat-stable skeletal muscle inhibitor protein of PKA. Peptide binds to the catalytic subunit of PKA and displaces the regulatory subunit. Mimics protein substrate by binding to the catalytic site via the Arg-cluster basic residues. Ki =2,3 nM for PKA.