Drukuj

7-Oxo-7H-benzo[de]benzo[4,5]imidazo[2,1-a]isoquinoline-3-carboxylic acid ≥98% (by TLC)

Dostawca: ENZO LIFE SCIENCES

Synonim: 7-Oxo-7H-benzo[de]benzo[4,5]imidazo[2,1-a]isoquinoline-3-carboxylic acid

BMLEI3890005 BMLEI3890025
ENZOBMLEI3890005EA 570 PLN
ENZOBMLEI3890005 ENZOBMLEI3890025
7-Oxo-7H-benzo[de]benzo[4,5]imidazo[2,1-a]isoquinoline-3-carboxylic acid ≥98% (by TLC)
STO-609

CaM kinase kinase inhibitor


STO-609 is a specific CaM kinase kinase (CaM-KK) inhibitor. Ki=0.2µM and 0.04µM for alpha and beta isotypes respectively. Demonstrates >80-fold selectivity for CAM-KK over CaMK1, CaMK2, CaMK4, MLCK, PKA, PKC, and p42 MAPK. Concentrations up to 10µg/ml do not affect cellular viability. It is a tool for exploring the role of the CaM-KK pathway in various physiological processes.

Formuła: C₁₉H₁₀N₂O₃
MW: 314.29 g/mol
Numer MDL: MFCD30685721
Numer CAS: 52029-86-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d98684ec65e0262f87caa444bd7643027398386&vid=a0be4941dd26648e1927390e3b0acee14b562885&prid=web&clid=SAPDAM

Zobacz ceny


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR