Drukuj

Anti-VASP Mouse Monoclonal Antibody

Dostawca: ENZO LIFE SCIENCES
ALX804240C100
ENZOALX804240C100EA 4230 PLN
ENZOALX804240C100
Anti-VASP Mouse Monoclonal Antibody
Przeciwciała
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat and rabbit Ser239-phosphorylated VASP. Does not cross-react with the non-phosphorylated form of VASP.

Reconstitute with 1ml water (15 minutes at room temperature).

Type: Primary
Antigen: VASP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
New Product
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR