Drukuj
Mouse recombinant BAFF (soluble)
Dostawca: ENZO LIFE SCIENCES
ALX522052C010
ENZOALX522052C010EA 2310 PLN
ENZOALX522052C010
Mouse recombinant BAFF (soluble)
Białka
Produced in HEK 293 cells. The extracellular domain of mouse BAFF (aa 127-309) is fused at the N-terminus to a linker peptide (10 aa) and a FLAG®-tag.