Drukuj

MGD sodium salt monohydrate 98 HPLC_ASSAY_METHOD

Dostawca: ENZO LIFE SCIENCES
Uwaga

Synonim: N-(Dithiocarbamoyl)-N-methyl-D-glucamine.Na.H₂O

ALX400014M050 ALX400014M250
ENZOALX400014M050EA 525 PLN
ENZOALX400014M050 ENZOALX400014M250
MGD sodium salt monohydrate 98 HPLC_ASSAY_METHOD
MGD sodium salt monohydrate

Spin trapping reagent.


Together with FeSO₄ MGD is a useful component for the formation of the MGD2-Fe²⁺ complex, which is an excellent nitric oxide (NO) spin-trapping reagent. The MGD2-Fe²⁺ complex is quite unstable, especially in the presence of dissolved oxygen. Thus, the complex should be used immediately after being made. An excess (usually 5-fold excess), of MGD to Fe²⁺ is used for making the complex with FeSO₄ to give a more stable complex solution. Acidic conditions should be avoided because dithiocarbamate tends to decompose forming toxic carbon disulfide.


Soluble in water.

Formuła: C₈H₁₆NO₅S₂Na·H₂O
MW: 311.3 g/mol
Temperatura przechowywania: Lodówka
Numer CAS: 91840-27-6

Zobacz ceny

Wyniki testów specyfikacyjnych

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR