Drukuj
Mouse recombinant TGF-beta3
Dostawca: ENZO LIFE SCIENCES
ALX2018170010 ALX2018170100
ENZOALX2018170010EA 1620 PLN
ENZOALX2018170010 ENZOALX2018170100
Mouse recombinant TGF-beta3
Białka
Produced in E. coli. Non-glycosylated, disulfide-linked homodimer, containing two 112 amino acid chains.
  • High purity
  • Carrier-free