Drukuj
Human recombinant Activin-A
Dostawca: ENZO LIFE SCIENCES
ALX2018090010 ALX2018090100
ENZOALX2018090010EA 1740 PLN
ENZOALX2018090010 ENZOALX2018090100
Human recombinant Activin-A
Białka
Produced in E. coli.
  • Carrier-free
Non-glycosylated homodimer, containing two 117 amino acid chains.