Drukuj
Zestaw ELISA do oznaczania HSF1
Dostawca: ENZO LIFE SCIENCES
ADI900198
ENZOADI900198EA 3850 PLN
ENZOADI900198
Zestaw ELISA do oznaczania HSF1
Zestawy do oznaczeń life science Testy ELISA
The HSF1 EIA kit is a complete kit for the quantitative determination of HSF1 in cell lysates and tissue extracts from human and mouse origins. This is a Sensitive ELISA kit for this key transcription factor for heat shock protein (chaperone) research.
  • Use less precious sample compared to Western blot
  • Ready to use liquid color-coded reagents and pre-coated plate reduces error
For the quantitative determination of human and mouse HSF1 in cell lysates and tissue.
Dodatkowe informacje: Supplied with antibody, assay buffer, conjugate, extraction reagent #1, microtiter plate, standard, stop solution, TMB substrate, wash buffer concentrate.