Drukuj
Zestaw ELISA do oznaczania IFN-gamma (człowiek)
Dostawca: ENZO LIFE SCIENCES
ADI900136
ENZOADI900136EA 4110 PLN
ENZOADI900136
Zestaw ELISA do oznaczania IFN-gamma (człowiek)
Zestawy do oznaczeń life science Testy ELISA
The IFN-gamma (Human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450 nm.
  • Ultrasensitive
  • Efficient - only 50 µL diluted sample needed per well
  • Measure natural and recombinant human IFN-γ
For the quantitative determination of human IFN-gamma in culture supernatants, plasma, serum, and urine.
Dodatkowe informacje: Supplied with microtiter plate, SA-HRP concentrate, SA-HRP dilution buffer, biotinylated antibody, standard, standard diluent, wash buffer concentrate, TMB substrate, stop solution