Drukuj
Dozownik do kapilar
Dostawca: CAMAG
022.7650 022.7655 022.7660 022.7661 022.7662 022.7665 022.7770 022.7771 022.7772 022.7775 022.7786
552-0062EA 4930 PLN
552-0062 552-0063 552-0064 552-0065 552-0066 552-0067 552-0068 552-0069 552-0070 552-0071 552-0044
Dozownik do kapilar
Kapilary do TLC
Compact capillary dispensers can be used on the Nanomat or independently. The capillary pipettes are loaded into the dispenser in magazines from a package of 100 capillaries. Capillaries of 0,5, 1, 2, and 5 µl volume are available in boxes of 500 capillaries (five belts).
  • Easy to use
  • All components are interchangeable from old to new systems
* With universal capillary holder (552-0044), a dispenser magazine for 1 µl capillaries (552-0065), and one box of 5×100 capillary pipettes of 1,0 µl (552-0069).