Drukuj
Komory z płaskim dnem
Dostawca: CAMAG
022.5150 022.5151 022.5250 022.5257 022.5259
552-0009EA 1140 PLN
552-0009 552-0010 552-0015 552-0022 552-0024
Komory z płaskim dnem
Komory do TLC
These flat bottom chamber allow the chromatogram to be developed in a tank atmosphere of partial or complete saturation with solvent vapours. The degree of layer pre-saturation cannot be controlled unless additional accessories are used.
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable