Drukuj
Jakościowe bibuły filtracyjne klasy 4, Whatman™
Dostawca: Whatman products (Cytiva)
1004-042 1004-047 1004-055 1004-070 1004-090 1004-110 1004-125 1004-150 1004-185 1004-240 1004-270 1004-320 1004-400 1004-917 1004-930 1004-027 1004-050 1004-648 1004-041
516-3402EA 56.9 PLN
516-3402 512-0412 512-1024 512-1025 512-1026 512-1027 512-1028 512-1029 512-1030 516-0601 512-0472 512-0384 512-0474 512-0385 512-0576 512-0470 512-0471 512-0643 WHAT1004-041
Jakościowe bibuły filtracyjne klasy 4, Whatman™
Filtry Sączki Sączki papierowe jakościowe
Whatman Grade 4 Qualitative Filter Papers are a standard grade, high flow rate filter paper. They are useful in qualitative analytical separations when fast flow is required and the collection of fine particles is not critical.
  • Excellent retention: Captures coarse particles and gelatinous precipitates such as ferric hydroxide and aluminum hydroxide
  • Fast filtration: Useful for routine clean-up of biological fluids or organic extracts during analysis
These cellulose filters are used in qualitative analytical techniques to determine and identifyWięcej informacji o produkcie