Drukuj
Jakościowe bibuły filtracyjne klasy 2, Whatman™
Dostawca: Whatman products (Cytiva)
1002-042 1002-047 1002-055 1002-070 1002-090 1002-110 1002-125 1002-185 1002-240 1002-270 1002-320 1002-917 1002-385 1002-929 1002-931 1002-147 1002-150
512-0381EA 64 PLN
512-0381 512-0574 512-1009 512-1010 512-1011 512-1012 512-1013 512-1015 516-0599 512-0458 512-0382 516-0600 512-0459 512-0462 512-0575 WHAT1002-147 512-1014
Jakościowe bibuły filtracyjne klasy 2, Whatman™
Filtry Sączki Sączki papierowe jakościowe
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
  • Each lot is guaranteed to meet the ASTM E 2187-09 specifications
  • Simplifies testing process by removing lot suitability testing
  • Just condition and use
Slightly more retentive than Grade 1 with a corresponding increase in filtration time (i.e., slightly slower filtration speed). More absorbent thanWięcej informacji o produkcie