Drukuj
Cut Protection Gloves, athletic C XP
Dostawca: UVEX
6003708 6003710
UVEX6003708EA 59.7 PLN
UVEX6003708 UVEX6003710
Cut Protection Gloves, athletic C XP
Rękawice
uvex athletic C XP offers very good cut protection with thumb reinforcement and breathable micro-foam coating.
  • 'Slim-fit' design and elastane in the liner
  • High breathability due to open-pore coating - sweating in the protective glove is reduced
  • Very good grip in dry and slightly damp areas
Upper material made of HPPE, glass, polyamide and elastane. Matte, open-pored and particularly abrasion-resistant microfoam coating. Additional thumb reinforcement ensures long service life and durability.
The uvex athletic C XP, with its comfortable 'slim-fit' design, enables flexible work with a high level of comfort and cut protection. The close, ergonomic fit allows for excellent feel and grip of workpieces.
The finely structured, open-pore micro foam coating makes the gloves highly breathable, reducing excessive sweating. The additional thumb reinforcement ensures increased durability, making the glove a particularly cost-effective product.
Application include Automotive industry, metal industry, glass industry, control work, sorting work.
Certyfikaty:
Certified according to EN 388 (4X43C).
EN ISO 21420:2020/EN 388:2016 + A1:2018; STANDARD 100 by OEKO-TEX®.