Drukuj
Mouse IgG1 ELISA Kits
Dostawca: Abcam
AB316264-10X96 AB316264-96TST
ABCAAB316264-10X96EA 38080 PLN
ABCAAB316264-10X96 ABCAAB316264-96TST
Mouse IgG1 ELISA Kits
Zestawy do oznaczeń life science Testy ELISA
Mouse IgG1 ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse IgG1 with a sensitivity of 40.7 ng/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Competitive ELISA - 450 nm readout: works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders .