Drukuj
Human IFN gamma ELISA Kit
Dostawca: Abcam
AB174443-96 AB174443-1X384
ABCAAB174443-96EA 3970 PLN
ABCAAB174443-96 ABCAAB174443-1X384
Human IFN gamma ELISA Kit
Zestawy do oznaczeń life science Testy ELISA
Human IFN gamma (IFNG) ELISA kit is a single-wash, 90-minute SimpleStep ELISA® a used to quantify human interferon-gamma (IFN-γ) with a sensitivity of 470 pg/ml. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. Colorimetric sandwich ELISA - 450 nm readout; compatible with any standard plate reader. Available in different formats to meet various throughput needs, including 384-well plates for higher throughput. Broad sample compatibility , suitable for serum, plasma (citrate, EDTA), and cell culture supernatants.