Drukuj
Mouse Thrombospondin 1 ELISA Kit
Dostawca: Abcam
AB317551-10X96 AB317551-1X384 AB317551-96TST
ABCAAB317551-10X96EA 36220 PLN
ABCAAB317551-10X96 ABCAAB317551-1X384 ABCAAB317551-96TST
Mouse Thrombospondin 1 ELISA Kit
Zestawy do oznaczeń life science Testy ELISA
Mouse Thrombospondin 1 ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse Thrombospondin 1 with a sensitivity of 0,054 ng/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.