Drukuj
Human Pro-Collagen I alpha 1 ELISA Kits
Dostawca: Abcam
AB210966-96 AB210966-1X384
ABCAAB210966-96EA 4310 PLN
ABCAAB210966-96 ABCAAB210966-1X384
Human Pro-Collagen I alpha 1 ELISA Kits
Zestawy do oznaczeń life science Testy ELISA
Human Pro-Collagen I alpha 1 ELISA kit is a single-wash, 90-minute SimpleStep ELISA® used to quantify human pro-collagen I alpha 1 with a sensitivity of 5.3 pg/ml. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. Colorimetric sandwich ELISA – 450 nm readout, compatible with any standard plate reader - Available in different formats – 384-well format available for high-throughput applications - Broad sample compatibility – works with serum, plasma, cell culture extracts, tissue extracts, and cell culture supernatant - Cited in over 40 publications.