Drukuj
Mouse FGF-7/KGF ELISA Kits
Dostawca: Abcam
AB317771-10X96 AB317771-1X384 AB317771-96TST
ABCAAB317771-10X96EA 0 PLN
ABCAAB317771-10X96 ABCAAB317771-1X384 ABCAAB317771-96TST
Mouse FGF-7/KGF ELISA Kits
Zestawy do oznaczeń life science Testy ELISA
Mouse FGF-7/KGF ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse FGF-7/KGF with a sensitivity of 5.036 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.