Mouse FGF-7/KGF ELISA Kits
Dostawca: Abcam
Mouse FGF-7/KGF ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse FGF-7/KGF with a sensitivity of 5.036 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.