Human PTH ELISA Kit
Dostawca: Abcam
Human PTH ELISA kit is a single-wash 90-min Simplestep used to quantify Human PTH with a sensitivity of 0,761 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders and 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 5 citations.