Drukuj
Monkey IL-6 ELISA Kit
Dostawca: Abcam
AB242233-1X96 AB242233-10X96 AB242233-1X384
ABCAAB242233-1X96EA 4200 PLN
ABCAAB242233-1X96 ABCAAB242233-10X96 ABCAAB242233-1X384
Monkey IL-6 ELISA Kit
Zestawy do oznaczeń life science Testy ELISA
Monkey IL-6 ELISA kit is a single-wash 90-min Simplestep used to quantify Monkey IL-6 with a sensitivity of 3,2 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.