Drukuj
Human AST ELISA Kit (Aspartate Aminotransferase)
Dostawca: Abcam
AB263881-1X96 AB263881-1X384
ABCAAB263881-1X96EA 4330 PLN
ABCAAB263881-1X96 ABCAAB263881-1X384
Human AST ELISA Kit (Aspartate Aminotransferase)
Zestawy do oznaczeń life science Testy ELISA
Human AST ELISA Kit (Aspartate Aminotransferase) is a single-wash 90-min Simplestep used to quantify Human AST (Aspartate Aminotransferase) with a sensitivity of 43 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput.