Drukuj
Human NOTUM ELISA Kit- Intracellular
Dostawca: Abcam
AB317780-10X96 AB317780-1X384 AB317780-96TST
ABCAAB317780-10X96EA 36220 PLN
ABCAAB317780-10X96 ABCAAB317780-1X384 ABCAAB317780-96TST
Human NOTUM ELISA Kit- Intracellular
Zestawy do oznaczeń life science Testy ELISA
Human NOTUM ELISA Kit- Intracellular is a single-wash 90-min Simplestep used to quantify Human NOTUM with a sensitivity of 17,047 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.