Human Lp-PLA2 ELISA Kits
Dostawca: Abcam
Human Lp-PLA2 ELISA kit is a single-wash 90-min Simplestep used to quantify Human Lp-PLA2 with a sensitivity of 45 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders.