Drukuj
Human IgE ELISA Kit
Dostawca: Abcam
AB317776-10X96 AB317776-1X384 AB317776-96TST
ABCAAB317776-10X96EA 35500 PLN
ABCAAB317776-10X96 ABCAAB317776-1X384 ABCAAB317776-96TST
Human IgE ELISA Kit
Zestawy do oznaczeń life science Testy ELISA
Human IgE ELISA kit is a single-wash 90-min Simplestep used to quantify Human IgE with a sensitivity of 23,1 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.