Drukuj
Anti-Cholera enterotoxin subunit B Rabbit Monoclonal Antibody [clone: EPR27372-4]
Dostawca: Abcam
AB317738-20UL AB317738-100UL AB317738-1ML
ABCAAB317738-20ULEA 1090 PLN
ABCAAB317738-20UL ABCAAB317738-100UL ABCAAB317738-1ML
Anti-Cholera enterotoxin subunit B Rabbit Monoclonal Antibody [clone: EPR27372-4]
Przeciwciała
Rabbit monoclonal [EPR27372-4] to Cholera enterotoxin subunit B.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Cholera enterotoxin subunit B
Clonality: Monoclonal
Clone: EPR27372-4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Zobacz ceny