Drukuj
Anti-RHEB Rabbit Monoclonal Antibody [clone: EPR25129-17]
Dostawca: Abcam
AB316265-20UL AB316265-100UL AB316265-1ML
ABCAAB316265-20ULEA 1090 PLN
ABCAAB316265-20UL ABCAAB316265-100UL ABCAAB316265-1ML
Anti-RHEB Rabbit Monoclonal Antibody [clone: EPR25129-17]
Przeciwciała
Rabbit monoclonal [EPR25129-17] to RHEB.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: RHEB
Clonality: Monoclonal
Clone: EPR25129-17
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat