Drukuj

Anti-ZCWCC1 Rabbit Monoclonal Antibody [clone: EPR28655-56]

Dostawca: Abcam
AB315904-1MG AB315904-100UG
ABCAAB315904-1MGEA 40660 PLN
ABCAAB315904-1MG ABCAAB315904-100UG
Anti-ZCWCC1 Rabbit Monoclonal Antibody [clone: EPR28655-56]
Przeciwciała
Rabbit monoclonal [EPR28655-56] to ZCWCC1 - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: ZCWCC1
Clonality: Monoclonal
Clone: EPR28655-56
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR