Drukuj

Anti-GPCR GPR48/LGR4 Rabbit Monoclonal Antibody [clone: EPR26351-175]

Dostawca: Abcam
AB321790-100UG AB321790-1MG
ABCAAB321790-100UGEA 4530 PLN
ABCAAB321790-100UG ABCAAB321790-1MG
Anti-GPCR GPR48/LGR4 Rabbit Monoclonal Antibody [clone: EPR26351-175]
Przeciwciała
Rabbit monoclonal [EPR26351-175] to GPCR GPR48/LGR4 - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GPCR GPR48/LGR4
Clonality: Monoclonal
Clone: EPR26351-175
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR