Drukuj

Anti-Alpha-synuclein (phospho Y39) Rabbit Monoclonal Antibody [clone: EPR25118-5]

Dostawca: Abcam
AB317366-20UL AB317366-100UL AB317366-1ML
ABCAAB317366-20ULEA 1090 PLN
ABCAAB317366-20UL ABCAAB317366-100UL ABCAAB317366-1ML
Anti-Alpha-synuclein (phospho Y39) Rabbit Monoclonal Antibody [clone: EPR25118-5]
Przeciwciała
Rabbit monoclonal [EPR25118-5] to Alpha-synuclein (phospho Y39).

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Alpha-synuclein
Clonality: Monoclonal
Clone: EPR25118-5
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR