Drukuj

Anti-Pericentrin Rabbit Monoclonal Antibody [clone: EPR21987] (Alexa Fluor® 555)

Dostawca: Abcam
AB319181-10UL AB319181-100UL
ABCAAB319181-10ULEA 1060 PLN
ABCAAB319181-10UL ABCAAB319181-100UL
Anti-Pericentrin Rabbit Monoclonal Antibody [clone: EPR21987] (Alexa Fluor® 555)
Przeciwciała
Alexa Fluor® 555 Rabbit monoclonal [EPR21987] to Pericentrin - Centrosome Marker.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Pericentrin
Clonality: Monoclonal
Clone: EPR21987
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR