Drukuj

Anti-Ly6g Rabbit Monoclonal Antibody [clone: EPR22909-135] (PE (Phycoerythrin))

Dostawca: Abcam
AB303605-10UL AB303605-100UL
ABCAAB303605-10ULEA 1060 PLN
ABCAAB303605-10UL ABCAAB303605-100UL
Anti-Ly6g Rabbit Monoclonal Antibody [clone: EPR22909-135] (PE (Phycoerythrin))
Przeciwciała
PE Rabbit monoclonal [EPR22909-135] to Ly6g.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Ly6g
Clonality: Monoclonal
Clone: EPR22909-135
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR