Drukuj

Anti-DPP4 Rabbit Monoclonal Antibody [clone: EPR18215] (Biotin)

Dostawca: Abcam
AB317766-10UL AB317766-100UL
ABCAAB317766-10ULEA 1060 PLN
ABCAAB317766-10UL ABCAAB317766-100UL
Anti-DPP4 Rabbit Monoclonal Antibody [clone: EPR18215] (Biotin)
Przeciwciała
Biotin Rabbit monoclonal [EPR18215] to DPP4.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: DPP4
Clonality: Monoclonal
Clone: EPR18215
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR