Drukuj

Anti-Milk Fat Globule 1 Rabbit Monoclonal Antibody [clone: EPR29114-57]

Dostawca: Abcam
AB319152-20UL AB319152-100UL AB319152-1ML
ABCAAB319152-20ULEA 1090 PLN
ABCAAB319152-20UL ABCAAB319152-100UL ABCAAB319152-1ML
Anti-Milk Fat Globule 1 Rabbit Monoclonal Antibody [clone: EPR29114-57]
Przeciwciała
Rabbit monoclonal [EPR29114-57] to Milk Fat Globule 1.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Milk Fat Globule 1
Clonality: Monoclonal
Clone: EPR29114-57
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR