Drukuj

Anti-CD300C Rabbit Monoclonal Antibody

Dostawca: Abcam
AB319996-20UL AB319996-100UL AB319996-1ML
ABCAAB319996-20ULEA 1090 PLN
ABCAAB319996-20UL ABCAAB319996-100UL ABCAAB319996-1ML
Anti-CD300C Rabbit Monoclonal Antibody
Przeciwciała
Rabbit monoclonal [EPR29236-56] to CD300C.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD300C
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR