Drukuj
Anti-Pseudomonas Outer membrane porin F Rabbit Monoclonal Antibody [clone: EPR28629-91]
Dostawca: Abcam
AB316973-20UL AB316973-100UL AB316973-1ML
ABCAAB316973-20ULEA 1090 PLN
ABCAAB316973-20UL ABCAAB316973-100UL ABCAAB316973-1ML
Anti-Pseudomonas Outer membrane porin F Rabbit Monoclonal Antibody [clone: EPR28629-91]
Przeciwciała
Rabbit monoclonal [EPR28629-91] to Pseudomonas Outer membrane porin F.
Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Pseudomonas Outer membrane porin F
Clonality: Monoclonal
Clone: EPR28629-91
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pseudomonas aeruginosa