Drukuj

Anti-DPP10 Rabbit Monoclonal Antibody

Dostawca: Abcam
AB316772-20UL AB316772-100UL AB316772-1ML
ABCAAB316772-20ULEA 1090 PLN
ABCAAB316772-20UL ABCAAB316772-100UL ABCAAB316772-1ML
Anti-DPP10 Rabbit Monoclonal Antibody
Przeciwciała
Rabbit monoclonal [EPR29237-246] to DPP10.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: DPP10
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR